Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ YBX2 Monoclonal Antibody (A2-B9-G6)

Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA534886
Description
Positive Control: MCF-7, mouse testis tissue.
YBX2 is a germ-cell specific member of the vertebrate Y box family of nucleic acid-binding proteins.
Specifications
| YBX2 | |
| Monoclonal | |
| 2 mg/mL | |
| PBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q9Y2T7 | |
| YBX2 | |
| Synthetic peptide within Human MSY2 aa 295-344. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| A2-B9-G6 | |
| Unconjugated | |
| YBX2 | |
| Contrin; CSDA3; Dbpc; DNA-binding protein C; FRGY2 homolog; germ cell specific Y-box binding protein; germ cell-specific Y-box-binding protein; MSY2; MSY2 homolog; RGD1305068; Y box binding protein 2; Y box protein 2; Y-box binding protein 2; Y-box protein MSY2; Y-box-binding protein 2; Ybx2 | |
| Mouse | |
| Protein G | |
| RUO | |
| 51087 | |
| -20°C, Avoid Freeze/Thaw Cycles, store in dark | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction