Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ YBX2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5111440
Description
Immunogen sequence: PGNPATAVSG TPAPPARSQA DKPVLAIQVL GTVKW.
YBX2 is a germ-cell specific member of the vertebrate Y box family of nucleic acid-binding proteins.
Specifications
| YBX2 | |
| Polyclonal | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9Y2T7 | |
| YBX2 | |
| Recombinant protein fragments (Protein Epitope Signature Tags, PrESTs) corresponding to a.a. 66-100 in Human YBX2. Recombinant protein control fragment (Product #RP-101090). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| YBX2 | |
| Contrin; CSDA3; Dbpc; DNA-binding protein C; FRGY2 homolog; germ cell specific Y-box binding protein; germ cell-specific Y-box-binding protein; MSY2; MSY2 homolog; RGD1305068; Y box binding protein 2; Y box protein 2; Y-box binding protein 2; Y-box protein MSY2; Y-box-binding protein 2; Ybx2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 51087 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction