Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ YBX2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5101278
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5101278 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5101278 Supplier Invitrogen™ Supplier No. PA5101278
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total YBOX2.

YBX2 is a germ-cell specific member of the vertebrate Y box family of nucleic acid-binding proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen YBX2
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene YBX2
Gene Accession No. Q9Y2T7, Q9Z2C8
Gene Alias Contrin; CSDA3; Dbpc; DNA-binding protein C; FRGY2 homolog; germ cell specific Y-box binding protein; germ cell-specific Y-box-binding protein; MSY2; MSY2 homolog; RGD1305068; Y box binding protein 2; Y box protein 2; Y-box binding protein 2; Y-box protein MSY2; Y-box-binding protein 2; Ybx2
Gene Symbols YBX2
Host Species Rabbit
Immunogen A synthesized peptide derived from human YBX2(Accession Q9Y2T7), corresponding to amino acid residues Q291-P341.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 303250, 51087, 53422
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.