Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ YDJC Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566359
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566359 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566359 Supplier Invitrogen™ Supplier No. PA566359
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RVLEGTLAGHT LTAELMAHPG YPSVPPTGGC GEGPDAFSCS WERLHELRVL TAPTLRAQLA QDGVQLCALD DLDSKRPGEE VPCEPTLEPF Highest antigen sequence identity to the following orthologs - mouse 79%, rat 79%.

Probably catalyzes the deacetylation of acetylated carbohydrates an important step in the degradation of oligosaccharides.
TRUSTED_SUSTAINABILITY

Specifications

Antigen YDJC
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene YDJC
Gene Accession No. A8MPS7
Gene Alias 1810015A11Rik; 4930521M19Rik; Carbohydrate deacetylase; UPF0249 protein; UPF0249 protein ydjC homolog; YDJC; YdjC homolog (bacterial)
Gene Symbols YDJC
Host Species Rabbit
Immunogen Recombinant Human YDJC. Recombinant protein control fragment (Product #RP-107038).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 150223
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.