Learn More
Thermo Scientific™ Sensititre™ YeastOne™ YO2IVD AST Plate
Catalog No. styo2ivd
Click to view available options
Quantity:
10/Pk.
This item is not returnable. View return policy
This item is not returnable. View return policy
Simplify yeast testing for Candida sp. and reduce cost per test using this dual isolate plate with Voriconazole.
Simplify yeast testing for Candida sp. and reduce cost per test using the dual isolate Thermo Scientific™ Sensititre™ YeastOne™ YO2IVD AST Plate with Voriconazole. Colorimetric Thermo Scientific™ alamarBlue™ agent provides reliable, easy and consistent endpoint determination with visual read option for efficient workflow and accurate results.
Simplify yeast testing for Candida sp. and reduce cost per test using the dual isolate Sensititre YeastOne YO2IVD AST Plate with Voriconazole.
  • Room-temperature storage eliminates inventory control concerns.
  • Individual packaging allows laboratory to test one plate at a time with no waste.
  • Inclusive on-scale QC ranges provide immediate quality assurance of testing methodology.
  • Cost-effective: the dual isolate plate allows two patient samples per plate reducing the cost per test.
  • 24-hour incubation time enables timely results for clinicians. 
Not all products are available for sale in all territories. Please inquire.
VersaTREK™ and Sensititre™ products are now part of the Thermo Scientific brand.
TRUSTED_SUSTAINABILITY
Specifications
0.5 McFarland Standard H2O (Part No. T3339)
Antimicrobial Agents Various
Broth Type YeastOne Broth (Part No. Y3462)
Description Sensititre YeastOne IVD Plate, with Voriconazole
Antibiotic Code Various
Product Type Microbiology Media
Test Type Yeast Testing
Incubation Conditions 35°C O2
Incubation Time 24-25 hr or 48-72 hr for Cryptococcus* (*For research use only. Not for use in diagnostic procedures.)
Isolates Per Plate 2
No. per Pack 10
Quantity 10/Pk.
Reading Options Manual
Transfer Volume 20µL
Volume Per Well 100 μL
Species Candida species
Show More Show Less
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.