Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

YEATS2 Polyclonal Antibody, Invitrogen™

Catalog No. PIPA561115
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561115 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561115 Supplier Thermo Scientific Supplier No. PA561115

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Rabbit Polyclonal Antibody

Immunogen sequence: GLLKVSEGSK TCDTMVFNHP AIKKFLESPS RSSSPANQRA ETPSANHSES DSLSQHNDFL SDKDNNSNMD IEERLSNNME QRPSRNTGRD T Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 87%.

The YEATS domain-containing protein 2 (YEATS2) is a nuclear protein that is phosphorylated upon DNA damage by ATM or ATR and contains 1 YEATS domain. The gene encoding YEATS2 resides on chromosome 3, which contains about 214 million bases encoding over 1, 100 genes. Notably, there is a chemokine receptor gene cluster and a variety of human cancer related loci on chromosome 3. Particular regions of the chromosome 3 short arm are deleted in many types of cancer cells. Key tumor suppressing genes on chromosome 3 encode apoptosis mediator RASSF1, cell migration regulator HYAL1 and angiogenesis suppressor SEMA3B.
TRUSTED_SUSTAINABILITY

Specifications

Antigen YEATS2
Applications Immunocytochemistry, Immunohistochemistry
Classification Polyclonal
Concentration 0.6 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene YEATS2
Gene Accession No. Q9ULM3
Gene Alias BC042768; Kiaa1197; mKIAA1197; YEATS domain containing 2; YEATS domain-containing protein 2; Yeats2; YETS2
Gene Symbols YEATS2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human YEATS2
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55689
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less

Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.