Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Yellow Fever Virus Antibody (0G5), Novus Biologicals™
SDP

Catalog No. NB10064510 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10064510 0.05 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10064510 Supplier Novus Biologicals Supplier No. NB10064510
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 2 publications

Yellow Fever Virus Monoclonal specifically detects Yellow Fever Virus in Virus samples. It is validated for Immunocytochemistry/Immunofluorescence, Functional.

Specifications

Antigen Yellow Fever Virus
Applications Immunocytochemistry, Immunofluorescence, Functional Assay
Classification Monoclonal
Clone 0G5
Conjugate Unconjugated
Dilution Immunocytochemistry/Immunofluorescence 1:10-1:2000, Functional Assay
Formulation Ascites with No Preservative
Gene Alias yellow fever virus
Host Species Mouse
Immunogen 17D strain of yellow virus
Purification Method Unpurified
Quantity 0.05 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Specific for the envelope protein of the wild (Asibi) and vaccine strains of yellow fever virus. The clone has been reported to have neutralizing activity against the Asibi strain. No cross reactivity with other flaviviruses has been reported.
Target Species Virus
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Ascites
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.