Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Yellow Fever Virus NS1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP313336
This item is not returnable.
View return policy
Description
Yellow Fever Virus NS1 Polyclonal antibody specifically detects Yellow Fever Virus NS1 in Virus samples. It is validated for Western BlotSpecifications
Yellow Fever Virus NS1 | |
Polyclonal | |
PBS, 20% Glycerol (pH7) | |
Recombinant protein encompassing a sequence within the C-terminus region of Yellow Fever Virus NS1 Protein (Yellow fever virus (strain 17D vaccine)). The exact sequence is proprietary. | |
100 μg | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Western Blot | |
Unconjugated | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Virus | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction