Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Yellow Fever Virus NS1 Monoclonal Antibody (3572)
GREENER_CHOICE

Catalog No. PIMA533300
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA533300 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA533300 Supplier Invitrogen™ Supplier No. MA533300
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Non-reactive in Tick-borne encephalitis virus, West Nile virus, and Japanese encephalitis virus.

Yellow fever virus is the pathogenic agent of the mosquito-borne Yellow fever disease. A member of the Flavivirus family, yellow fever virus is enveloped and contains an RNA genome. The virus causes disease by replicating in lymph nodes and infecting dendritic cells and hepatocytes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Yellow Fever Virus NS1
Applications ELISA
Classification Monoclonal
Clone 3572
Concentration 100 μg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.2
Gene Alias yellow fever virus
Host Species Mouse
Purification Method Protein A
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.