Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Yellow Fever Virus NS1 Monoclonal Antibody (3573)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA533301
Description
Non-reactive in Tick-borne encephalitis virus, West Nile virus, and Japanese encephalitis virus.
Yellow fever virus is the pathogenic agent of the mosquito-borne Yellow fever disease. A member of the Flavivirus family, yellow fever virus is enveloped and contains an RNA genome. The virus causes disease by replicating in lymph nodes and infecting dendritic cells and hepatocytes.
Specifications
Yellow Fever Virus NS1 | |
Monoclonal | |
100 μg/mL | |
PBS with 0.1% sodium azide; pH 7.2 | |
Mouse | |
1 mL | |
Primary | |
4°C | |
Liquid |
ELISA | |
3573 | |
Unconjugated | |
yellow fever virus | |
Protein A | |
RUO | |
Virus | |
Antibody | |
IgG1 |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction