Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Yellow Fever Virus NS1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5112026
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5112026 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5112026 Supplier Invitrogen™ Supplier No. PA5112026
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Yellow Fever Virus NS1 Polyclonal Antibody for Western Blot

Yellow fever virus is the pathogenic agent of the mosquito-borne Yellow fever disease. A member of the Flavivirus family, yellow fever virus is enveloped and contains an RNA genome. The virus causes disease by replicating in lymph nodes and infecting dendritic cells and hepatocytes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Yellow Fever Virus NS1
Applications Western Blot
Classification Polyclonal
Concentration 2.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gene Alias yellow fever virus
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the C-terminus region of Yellow Fever Virus NS1 Protein (Yellow fever virus (strain 17D vaccine)). The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.