Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Yellow Fever Virus NS2B Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP313337
This item is not returnable.
View return policy
Description
Yellow Fever Virus NS2B Polyclonal antibody specifically detects Yellow Fever Virus NS2B in Virus samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
| Yellow Fever Virus NS2B | |
| Polyclonal | |
| PBS, 20% Glycerol (pH7) | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| Virus | |
| Purified |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| Flavivirin protease NS2B regulatory subunit, Non-structural protein 2B, Serine protease subunit NS2B, YFV NS2B | |
| Full length Yellow Fever Virus NS2B recombinant protein. (Yellow fever virus (strain 17D vaccine)) | |
| 100 μg | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction