Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Yellow Fever Virus NS3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP313322
This item is not returnable.
View return policy
Description
Yellow Fever Virus NS3 Polyclonal antibody specifically detects Yellow Fever Virus NS3 in Virus samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
| Yellow Fever Virus NS3 | |
| Polyclonal | |
| PBS, 20% Glycerol (pH7) | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| Virus | |
| Purified |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| Flavivirin protease NS3 catalytic subunit, Non-structural protein 3, Serine protease NS3, YFV NS3 | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the center region of Yellow Fever Virus NS3 protein (Yellow fever virus (strain 17D vaccine)). The exact sequence is proprietary. | |
| 100 μg | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction