Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Yellow Fever Virus NS3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5112021
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5112021 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5112021 Supplier Invitrogen™ Supplier No. PA5112021
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Yellow Fever Virus NS3 Polyclonal Antibody for Western Blot, ICC/IF

Yellow fever virus (YFV) is an RNA virus belonging to the genus Flavivirus in the family Flaviviridae. YFV measures 40-60 nm in diameter and has a spherical, enveloped structure containing a single-stranded positive-sense RNA genome. The genome encodes three structural proteins (C, prM/M, and E) and seven nonstructural proteins (NS1, NS2A, NS2B, NS3, NS4A, NS4B, and NS5). YFV is transmitted by Aedes mosquitoes and is endemic in tropical regions of Africa and South America. It causes yellow fever, a hemorrhagic disease characterized by symptoms such as fever, jaundice, hemorrhage, and organ failure. Pathogenesis involves liver infection leading to hepatocyte and Kupffer cell damage, with the virus utilizing specific genes and molecular mechanisms for neurotropism and viscerotropism.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Yellow Fever Virus NS3
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 2.44 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Host Species Rabbit
Immunogen Carrier-protein conjugated synthetic peptide encompassing a sequence within the center region of Yellow Fever Virus NS3 protein (Yellow fever virus (strain 17D vaccine)). The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.