Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Yellow Wax, Cake, NF, Spectrum™ Chemical

Click to view available options
Quantity:
5 lb.
12 kg
Packaging:
Poly Pail
Description
Spectrum™ Chemical Yellow Wax, Cake, NF - All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
Specifications
CAS Max % | 1 |
Melting Range | 62-65°C |
Packaging | Poly Pail |
Quantity | 12 kg |
Grade | NF |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction