Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Yersinia pestis V Antigen Monoclonal Antibody (Va48), Invitrogen™

Catalog No. PIMA123088
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA123088 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA123088 Supplier Thermo Scientific Supplier No. MA123088

Mouse Monoclonal Antibody

MA1-23088 detects Yersinia pestis v antigen in Bacteria samples.

Yersinia pestis V antigen (LcrV) is an environmentally regulated, plasmid pCD1-encoded protein. Yersinia pestis is the etiologic agent of plague, and LcrV is a multifunctional protein that has been shown to act at the level of secretion control by binding the Ysc inner-gate protein LcrG and to modulate the host immune response by altering cytokine production. LcrV also is essential for the unidirectional targeting of Yops to the cytosol of infected eukaryotic cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Yersinia pestis V Antigen
Applications ELISA, Western Blot
Classification Monoclonal
Clone Va48
Concentration 4 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene Alias Bubonic Plague; Pasturella pestis; yersinia pseudotuberculosis
Host Species Mouse
Immunogen Recombinant full length protein.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.