Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

YES1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501985943
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-5943 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-5943 Supplier Bioss Supplier No. BS4166R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

YES is a member of the Src family of non-receptor tyrosine kinases participating in a wide range of cellular signalling pathways. The gene that encodes YES is the cellular homolog of the Yamaguchi sarcoma virus oncogene. This gene lies in proximity to thymidylate synthase gene on chromosome 18, and a pseudogene of it has mapped to chromosome 22.

Specifications

Antigen YES1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Yes1
Gene Accession No. P07947
Gene Alias cellular yes-1 protein; c-Src; c-yes; EC 2.7.10.2; HsT441; p60c-yes; p60-Src; p61-Yes; pp60c-src; Proto-oncogene c-Yes; proto-oncogene tyrosine-protein kinase YES; Tyrosine-protein kinase Yes; viral oncogene yes homolog; v-yes-1 Yamaguchi sarcoma viral oncogene homolog 1; Yamagichi sarcoma viral (v-yes-1) oncogene homolog 1; Yamaguchi sarcoma oncogene; Yamaguchi sarcoma viral (v-yes) oncogene homolog 1; Yamaguchi sarcoma viral oncogene homolog 1; Yes; YES proto-oncogene 1, Src family tyrosine kinase; Yes1; YES1 proto-oncogene, Src family tyrosine kinase
Gene Symbols Yes1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human YES1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7525
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.