Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
YES1 Rabbit anti-Mouse, Rat, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA572233
Description
YES is a member of the Src family of non-receptor tyrosine kinases participating in a wide range of cellular signalling pathways. The gene that encodes YES is the cellular homolog of the Yamaguchi sarcoma virus oncogene. This gene lies in proximity to thymidylate synthase gene on chromosome 18, and a pseudogene of it has mapped to chromosome 22.
Specifications
YES1 | |
Polyclonal | |
Unconjugated | |
Yes1 | |
HsT441; P61-YES; Yes; c-yes; Yamaguchi sarcoma oncogene; cellular yes-1 protein; proto-oncogene c-Yes; proto-oncogene tyrosine-protein kinase YES; tyrosine-protein kinase Yes | |
Rabbit | |
Protein A | |
RUO | |
22612, 24884 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.68 mg/mL | |
PBS with 0.09% sodium azide | |
F1LM93, Q04736 | |
Yes1 | |
KLH conjugated synthetic peptide between 148-182 amino acids from the Central region of Mouse Yes1 | |
400 μL | |
Primary | |
Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction