Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ YIF1A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5140186
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5140186 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5140186 Supplier Invitrogen™ Supplier No. PA5140186
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: TGADVAFSVN HLLGDPMANV AMAYGSSIAS HGKDMVHKEL HRFVSVSKLK Y Highest antigen sequence indentity to the following orthologs: Rat - 82%, Mouse - 84%.

YIF1A has a possible role in transport between endoplasmic reticulum and Golgi.
TRUSTED_SUSTAINABILITY

Specifications

Antigen YIF1A
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene YIF1A
Gene Accession No. O95070
Gene Alias 5330422J23Rik; 54TM; 54TMp; FinGER7; HYIF1P; liver protein; protein YIF1A; putative Rab5-interacting protein; putative transmembrane protein 54TMp; putative transmembrane protein; homolog of yeast Golgi membrane protein Yif1p (Yip1p-interacting factor); Yif1; YIF1A; Yif1p; Yip1 interacting factor homolog A; Yip1 interacting factor homolog A (S. cerevisiae); Yip1 interacting factor homolog A, membrane trafficking protein; YIP1-interacting factor homolog A; Yip1p-interacting factor
Gene Symbols YIF1A
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human YIF1A. Recombinant protein control fragment (Product #RP-109541).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10897
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.