Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ YIF1A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA545929
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA545929 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA545929 Supplier Invitrogen™ Supplier No. PA545929
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: SDGYYVALAW TSSALMYFIV RSLRTAALGP DSMGGPVPRQ RLQLYLTLGA.

YIF1A has a possible role in transport between endoplasmic reticulum and Golgi.
TRUSTED_SUSTAINABILITY

Specifications

Antigen YIF1A
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene YIF1A
Gene Accession No. O95070
Gene Alias 5330422J23Rik; 54TM; 54TMp; FinGER7; HYIF1P; liver protein; protein YIF1A; putative Rab5-interacting protein; putative transmembrane protein 54TMp; putative transmembrane protein; homolog of yeast Golgi membrane protein Yif1p (Yip1p-interacting factor); Yif1; YIF1A; Yif1p; Yip1 interacting factor homolog A; Yip1 interacting factor homolog A (S. cerevisiae); Yip1 interacting factor homolog A, membrane trafficking protein; YIP1-interacting factor homolog A; Yip1p-interacting factor
Gene Symbols YIF1A
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human YIF1A (aa 227-276).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10897
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.