Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
YIF1B Antibody, Alexa Fluor™ 405, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP318478AF405
Description
YIF1B Polyclonal antibody specifically detects YIF1B in Human samples. It is validated for Western BlotSpecifications
YIF1B | |
Polyclonal | |
Western Blot | |
FinGER8, FLJ23477, FLJ50074, MGC39135, protein YIF1B, Yip1 interacting factor homolog B (S. cerevisiae), YIP1-interacting factor homolog B | |
Recognizes region between residues Between between 1 and 50.. | |
0.1 mL | |
Neuroscience | |
90522 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
Alexa Fluor 405 | |
50mM Sodium Borate | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction