Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

YIF1B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA542659
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA542659 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA542659 Supplier Thermo Scientific Supplier No. PA542659
Add to Cart
Add to Cart

YIF1B Polyclonal Antibody

YIF1B Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen YIF1B
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene YIF1B
Gene Accession No. Q5BJH7
Gene Alias 9430029K10Rik; FinGER8; PP4519; Protein YIF1B; UNQ3073/PRO9905; YIF1B; Yip1 interacting factor homolog B; Yip1 interacting factor homolog B (S. cerevisiae); Yip1 interacting factor homolog B, membrane trafficking protein; Yip1b; YIP1-interacting factor homolog B
Gene Symbols YIF1B
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human YIF1B.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 90522
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.