Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ YIL 781 hydrochloride
GREENER_CHOICE

Catalog No. 395925
Change view
Click to view available options
Quantity:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
39-592-5 25 mg
39-595 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 39-592-5 Supplier Tocris Bioscience™ Supplier No. 3959/25
Add to Cart
Add to Cart
This item is not returnable. View return policy

Ghrelin receptor (GHS-R1a) antagonist

YIL 781 hydrochloride is a ghrelin receptor antagonist (GHS-R1a) (Ki = 17 nM). Displays no significant affinity for the motilin receptor (Ki = 6 μM). Blocks the effects of ghrelin on insulin secretion both in vivo and in vitro. Improves glucose homeostasis in vivo.

Specifications

Chemical Name or Material 6-(4-Fluorophenoxy)-2-methyl-3-[[(3S)-1-(1-methylethyl)-3-piperidinyl]methyl]-4(3H)-quinazolinone hydrochloride
CAS 875258-85-8
Assay Percent Range >98%
Quantity 25 mg
Molecular Formula C24H28FN3O2.HCl
Formula Weight Observed MW: 445.96
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.