Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
YIPF1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $487.50
Specifications
Antigen | YIPF1 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP17075020
![]() |
Novus Biologicals
NBP17075020UL |
20 μL |
Each for $206.00
|
|
|||||
NBP170750
![]() |
Novus Biologicals
NBP170750 |
100 μL |
Each for $487.50
|
|
|||||
Description
YIPF1 Polyclonal specifically detects YIPF1 in Human samples. It is validated for Western Blot.Specifications
YIPF1 | |
Polyclonal | |
Rabbit | |
Yip1 domain family, member 1 | |
YIPF1 | |
IgG |
Western Blot | |
Unconjugated | |
RUO | |
54432 | |
Synthetic peptides corresponding to YIPF1(Yip1 domain family, member 1) The peptide sequence was selected from the middle region of YIPF1. Peptide sequence HLGEKTYHYVPEFRKVSIAATIIYAYAWLVPLALWGFLMWRNSKVMNIVS. | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title