Learn More
Invitrogen™ YIPF3 Polyclonal Antibody

Description
Immunogen sequence: ARNGAGPEWGG FEENIQGGGS AVIDMENMDD TSGSSFEDMG ELHQRLREEE VDADAADAAA AEEEDGEFLG M Highest antigen sequence identity to the following orthologs - mouse 94%, rat 94%.
Specifications
Specifications
| Antigen | YIPF3 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | YIPF3 |
| Gene Accession No. | Q9GZM5 |
| Gene Alias | C6orf109; D17Wsu94e; dJ337H4.3; FinGER3; Killer lineage protein 1; KLIP1; Liver regeneration-related protein LRRGT00110; LRRGT00110; natural killer cell-specific antigen KLIP1; Protein YIPF3; Protein YIPF3, 36 kDa form III; Protein YIPF3, N-terminally processed; RGD1308185; Yip1 domain family member 3; Yip1 domain family, member 3; YIP1 family member 3; YIPF3 |
| Gene Symbols | YIPF3 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.