Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ YIPF3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113335
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113335 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113335 Supplier Invitrogen™ Supplier No. PA5113335
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

YIPF3 Polyclonal Antibody for Western Blot, ELISA

Involved in the maintenance of the Golgi structure. May play a role in hematopoiesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen YIPF3
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene YIPF3
Gene Accession No. Q3UDR8, Q9GZM5
Gene Alias C6orf109; D17Wsu94e; dJ337H4.3; FinGER3; Killer lineage protein 1; KLIP1; Liver regeneration-related protein LRRGT00110; LRRGT00110; natural killer cell-specific antigen KLIP1; Protein YIPF3; Protein YIPF3, 36 kDa form III; Protein YIPF3, N-terminally processed; RGD1308185; Yip1 domain family member 3; Yip1 domain family, member 3; YIP1 family member 3; YIPF3
Gene Symbols YIPF3
Host Species Rabbit
Immunogen Recombinant Human Protein YIPF3 protein (2-148AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25844, 28064
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.