Learn More
Invitrogen™ YIPF4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA521786
Description
Recommended positive controls: 293T, A431, H1299, HeLaS3, HepG2, Molt-4, Raji. Predicted reactivity: Mouse (97%), Rat (97%), Zebrafish (84%), Xenopus laevis (94%), Chicken (95%), Rhesus Monkey (100%), Chimpanzee (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Involved in the maintenance of the Golgi structure.
Specifications
YIPF4 | |
Polyclonal | |
Unconjugated | |
YIPF4 | |
2310034L04Rik; FinGER4; Nbla11189; Protein YIPF4; putative protein product of Nbla11189; RGD1310157; Yip1 domain family member 4; Yip1 domain family, member 4; YIP1 family member 4; Yipf4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
84272 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
Q9BSR8 | |
YIPF4 | |
Recombinant protein encompassing a sequence within the center region of human YIPF4. The exact sequence is proprietary. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.