Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ YIPF4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553656
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553656 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553656 Supplier Invitrogen™ Supplier No. PA553656
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PTNGDFTFVS SADAEDLSGS IASPDVKLNL GGDFIKESTA TTFLRQRGYG WLLEVEDDDP EDNKPLLEEL DIDLKDIYYK IRCVLMPMPS LGFN Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 95%.

Involved in the maintenance of the Golgi structure.
TRUSTED_SUSTAINABILITY

Specifications

Antigen YIPF4
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene YIPF4
Gene Accession No. Q9BSR8
Gene Alias 2310034L04Rik; FinGER4; Nbla11189; Protein YIPF4; putative protein product of Nbla11189; RGD1310157; Yip1 domain family member 4; Yip1 domain family, member 4; YIP1 family member 4; Yipf4
Gene Symbols YIPF4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human YIPF4. Recombinant protein control fragment (Product #RP-91227).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84272
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.