Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ YIPF5 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5113212
Description
YIPF5 Polyclonal Antibody for Western Blot, ICC/IF, ELISA
Plays a role in transport between endoplasmic reticulum and Golgi.
Specifications
YIPF5 | |
Polyclonal | |
Unconjugated | |
Yipf5 | |
2610311I19Rik; AA408236; Ac2-256; FinGER5; Five-pass transmembrane protein localizing in the Golgi apparatus and the endoplasmic reticulum 5; golgi membrane protein SB140; PP12723; Protein YIPF5; protein YIPF5-like protein; SB140; SMAP5; SMAP-5; smooth muscle cell associated protein 5; smooth muscle cell-associated protein 5; UNQ3123/PRO10275; Yip1 domain family member 5; Yip1 domain family, member 5; YIP1 family member 5; Yip1a; Yipf5; YPT-interacting protein 1 A | |
Rabbit | |
Protein G | |
RUO | |
67180, 81555 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
Q969M3, Q9EQQ2 | |
Yipf5 | |
Recombinant Human Protein YIPF5 protein (1-124AA). | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction