Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
YIPF5 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA567301
Description
Immunogen sequence: SGFENLNTDFY QTSYSIDDQS QQSYDYGGSG GPYSKQYAGY DYSQQGRFVP PD Highest antigen sequence identity to the following orthologs - mouse 91%, rat 89%.
Plays a role in transport between endoplasmic reticulum and Golgi.
Specifications
YIPF5 | |
Polyclonal | |
Unconjugated | |
Yipf5 | |
2610311I19Rik; AA408236; Ac2-256; FinGER5; Five-pass transmembrane protein localizing in the Golgi apparatus and the endoplasmic reticulum 5; golgi membrane protein SB140; PP12723; Protein YIPF5; protein YIPF5-like protein; SB140; SMAP5; SMAP-5; smooth muscle cell associated protein 5; smooth muscle cell-associated protein 5; UNQ3123/PRO10275; Yip1 domain family member 5; Yip1 domain family, member 5; YIP1 family member 5; Yip1a; Yipf5; YPT-interacting protein 1 A | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
81555 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry, Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q969M3 | |
Yipf5 | |
Recombinant Human YIPF5 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction