Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ YIPF7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA595713
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA595713 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA595713 Supplier Invitrogen™ Supplier No. PA595713
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MDLLKISHTK LHLLEDLSIK NKQRMSNLAQ FDSDFYQSNF TIDNQEQSGN DSNAYGNLYG SRKQQAGEQP QPASFVPSEM; Positive Samples: Rat heart, Rat liver; Cellular Location: Endoplasmic reticulum membrane, Multi-pass membrane protein.

YIPF7 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen YIPF7
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.96 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene YIPF7
Gene Alias FinGER9; Five-pass transmembrane protein localizing in the Golgi apparatus and the endoplasmic reticulum 9; protein YIPF7; RGD1310766; Yip1 domain family member 7; Yip1 domain family, member 7; YIP1 family member 7; YIP1B; YIPF7
Gene Symbols YIPF7
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-80 of human YIPF7 (NP_8723982).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 364147
Target Species Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.