Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ YIPF7 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA556528
Description
Immunogen sequence: LTVLNPMKPV DGSIMNETDL TGPILFCVAL GATLLLAG Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 95%.
YIPF7 is a protein coding gene.
Specifications
YIPF7 | |
Polyclonal | |
Unconjugated | |
YIPF7 | |
FinGER9; Five-pass transmembrane protein localizing in the Golgi apparatus and the endoplasmic reticulum 9; protein YIPF7; RGD1310766; Yip1 domain family member 7; Yip1 domain family, member 7; YIP1 family member 7; YIP1B; YIPF7 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
285525 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8N8F6 | |
YIPF7 | |
Recombinant protein corresponding to Human YIPF7. Recombinant protein control fragment (Product #RP-102348). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction