Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ YKT6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA595880
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA595880 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA595880 Supplier Invitrogen™ Supplier No. PA595880
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EFSKQVDRID WPVGSPATIH YPALDGHLSR YQNPREADPM TKVQAELDET KIILHNTMES LLERGEKLDD LVSKSEVLGT QSKAFYKTAR KQNSCCAIM.

This gene product is one of the SNARE recognition molecules implicated in vesicular transport between secretory compartments. It is a membrane associated, isoprenylated protein that functions at the endoplasmic reticulum-Golgi transport step. This protein is highly conserved from yeast to human and can functionally complement the loss of the yeast homolog in the yeast secretory pathway.
TRUSTED_SUSTAINABILITY

Specifications

Antigen YKT6
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.95 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene YKT6
Gene Accession No. O15498
Gene Alias 0610042I15Rik; 1810013M05Rik; AW105923; prenylated SNARE protein; prenylated SNARE protein (Ykt6); R-SNARE; SNARE protein Ykt6; synaptobrevin homolog YKT6; Ykt6; YKT6 homolog; YKT6 homolog (S. Cerevisiae); YKT6 v-SNARE homolog (S. cerevisiae); YKT6 v-SNARE protein; YKT6, S. cerevisiae, homolog of
Gene Symbols YKT6
Host Species Rabbit
Immunogen Synthetic peptide.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10652
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.