Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

YL-1 Mouse anti-Mouse, Human, Clone: EX-1Y32, MilliporeSigma™

Catalog No. MABE179625 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABE1796-25 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABE1796-25 Supplier MilliporeSigma Supplier No. MABE179625UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-YL-1, clone EX-1Y32 is a highly specific mouse monoclonal antibody, that targets Vacuolar protein sorting-associated protein 72 homolog and has been tested in Immunocytochemistry, Western Blotting and Immunoprecipitation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen YL-1
Applications Immunocytochemistry, Western Blot, Immunoprecipitation
Classification Monoclonal
Clone EX-1Y32
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols VPS72,TCFL1,YL1
Host Species Mouse
Immunogen GST-tagged full-length recombinant human YL-1 protein.
Purification Method Protein A + Protein G
Quantity 25 μg
Regulatory Status RUO
Research Discipline Epigenetics and Nuclear Function
Primary or Secondary Primary
Test Specificity Clone EX-1Y32 detects Vacuolar protein sorting-associated protein 72 homolog (YL-1) in human and murine cells.
Target Species Mouse, Human
Content And Storage Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.