Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
YL1 Antibody, PerCP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP171870PCP
Description
YL1 Polyclonal antibody specifically detects YL1 in Human samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
YL1 | |
Polyclonal | |
Western Blot, Immunoprecipitation | |
Protein YL-1, Swc2, TCFL1, Transcription factor-like 1, transcription factor-like 1 (YL-1), vacuolar protein sorting 72 (yeast), vacuolar protein sorting 72 homolog (S. cerevisiae), vacuolar protein sorting-associated protein 72 homolog, YL-1, YL1CFL1 | |
The immunogen for this product maps to a region between residue 50 and 100 of human Protein YL-1 using the numbering given in entry NP_005988.1 (GeneID 6944). | |
0.1 mL | |
Autophagy | |
6944 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunoprecipitation | |
PerCP | |
PBS | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction