Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ YM 022
GREENER_CHOICESDP

Catalog No. 140850 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-085-0 50 mg
14-081-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 14-085-0 Supplier Tocris Bioscience™ Supplier No. 1408/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

Highly potent, selective non-peptide CCK2 antagonist

Extremely potent and highly selective non-peptide CCK2 silent antagonist (Ki values are 68 pM and 63 nM at CCK2 and CCK1 receptors respectively). Acts in vivo to potently inhibit gastrin-induced gastric acid secretion and histidine decarboxylase activation with a long duration of action.

Chemical Identifiers

CAS 145084-28-2
Molecular Formula C32H28N4O3

Specifications

Quantity 50 mg
Formula Weight 516.6
Purity >99%
Molecular Weight (g/mol) 516.601
Chemical Name or Material YM 022
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.