Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ YM 155
GREENER_CHOICESDP

Catalog No. 649150 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
64-915-0 50 mg
64-911-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 64-915-0 Supplier R&D Systems™ Supplier No. 6491/50
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Survivin suppressor

YM 155 is a survivin suppressor. Down-regulates survivin expression and sensitizes human NSCLC cell lines to radiation. Enhances radiation-induced tumor regression in nude mice bearing NSCLC xenografts. Also induces apoptosis in TRAIL-resistant glioma cells in vitro.

Specifications

Chemical Name or Material 4,9-Dihydro-1-(2-methoxyethyl)-2-methyl-4,9-dioxo-3-(2-pyrazinylmethyl)-1H-naphth[2,3-d]imidazolium bromide
CAS 781661-94-7
Quantity 50 mg
Product Line Tocris
Molecular Formula C20H19BrN4O3
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.