Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ YM 58483
GREENER_CHOICE

Catalog No. 393910
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
39-391-0 10 mg
39-395-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 39-391-0 Supplier Tocris Bioscience™ Supplier No. 3939/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Inhibitor of SOCE in non-excitable cells

YM 58483 is a blocker of store-operated Ca2+ entry (SOCE), which mediates the activation of non-excitable cells (e.g. lymphocytes). Inhibits calcium release-activated calcium (CRAC) channels; suppresses thapsigargin -induced sustained Ca2+ influx (IC50 = 100 nM). Displays immuno-modulatory and anti-inflammatory effects; suppresses cytokine production and proliferation of T cells in vitro.

Chemical Identifiers

CAS 223499-30-7
Molecular Formula C15H9F6N5OS

Specifications

Quantity 10 mg
Formula Weight 421.32
Purity >99%
Molecular Weight (g/mol) 421.321
Chemical Name or Material YM 58483
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.