Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

YME1L1 Mouse anti-Human, Rat, Clone: 2B7E4, Proteintech
SDP

Catalog No. 501736971 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-6971 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-6971 Supplier Proteintech Group Inc Supplier No. 665511IG150UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

YMEL1 is the human ortholog of yeast mitochondrial AAA metalloprotease, Yme1p. It is localized in the mitochondria and can functionally complement a yme1 disruptant yeast strain. It is proposed that this gene plays a role in mitochondrial protein metabolism and could be involved in mitochondrial pathologies.

Specifications

Antigen YME1L1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 2B7E4
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene YME1L1
Gene Accession No. Q925S8, Q96TA2
Gene Alias FTSH, FTSH1, Meg 4, MEG4, PAMP, YME1 like 1 (S. cerevisiae), YME1 like protein 1, YME1L, YME1L1
Gene Symbols Yme1l1
Host Species Mouse
Immunogen YME1L1 Fusion Protein Ag2069
Purification Method Protein G
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10730, 114217
Target Species Human, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.