Learn More
Invitrogen™ YME1L1 Polyclonal Antibody

Description
Peptide sequence: SDFDNATKIA KRMVTKFGMS EKLGVMTYSD TGKLSPETQS AIEQEIRILL Sequence homology: Cow: 100%; Dog: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%.
Specifications
Specifications
| Antigen | YME1L1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 2% sucrose and 0.09% sodium azide |
| Gene | YME1L1 |
| Gene Accession No. | Q96TA2 |
| Gene Alias | ATP-dependent metalloprotease FtsH1; ATP-dependent metalloprotease FtsH1 homolog; ATP-dependent metalloprotease YME1L1; ATP-dependent metalloprotease YME1L1-like protein; ATP-dependent zinc metalloprotease YME1L1; ATP-dependent zinc metalloprotease YME1l1-like protein; FTSH; FTSH1; MEG4; meg-4; PAMP; Presenilin-associated metalloprotease; UNQ1868/PRO4304; YME1 like 1 ATPase; YME1L; YME1L1; YME1-like 1 (S. cerevisiae); YME1-like 1 ATPase; YME1-like protein 1 |
| Gene Symbols | YME1L1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.