Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ YME1L1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA564299
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564299 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564299 Supplier Invitrogen™ Supplier No. PA564299
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AVDGKEMVTM KELEFSKDKI LMGPERRSVE IDNKNKTITA YHESGHAIIA YYTKDAMPIN KATIMPRGPT LGHVSLLPEN DRWNETRAQL LAQMDVS Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.

YMEL1 is the human ortholog of yeast mitochondrial AAA metalloprotease, Yme1p. It is localized in the mitochondria and can functionally complement a yme1 disruptant yeast strain. It is proposed that this gene plays a role in mitochondrial protein metabolism and could be involved in mitochondrial pathologies.
TRUSTED_SUSTAINABILITY

Specifications

Antigen YME1L1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene YME1L1
Gene Accession No. Q96TA2
Gene Alias ATP-dependent metalloprotease FtsH1; ATP-dependent metalloprotease FtsH1 homolog; ATP-dependent metalloprotease YME1L1; ATP-dependent metalloprotease YME1L1-like protein; ATP-dependent zinc metalloprotease YME1L1; ATP-dependent zinc metalloprotease YME1l1-like protein; FTSH; FTSH1; MEG4; meg-4; PAMP; Presenilin-associated metalloprotease; UNQ1868/PRO4304; YME1 like 1 ATPase; YME1L; YME1L1; YME1-like 1 (S. cerevisiae); YME1-like 1 ATPase; YME1-like protein 1
Gene Symbols YME1L1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human YME1L1. Recombinant protein control fragment (Product #RP-103788).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10730
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.