Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ YME1L1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598779
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598779 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598779 Supplier Invitrogen™ Supplier No. PA598779
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

YME1L1 Polyclonal Antibody for Western Blot, IHC (P), ELISA

YMEL1 is the human ortholog of yeast mitochondrial AAA metalloprotease, Yme1p. It is localized in the mitochondria and can functionally complement a yme1 disruptant yeast strain. It is proposed that this gene plays a role in mitochondrial protein metabolism and could be involved in mitochondrial pathologies.
TRUSTED_SUSTAINABILITY

Specifications

Antigen YME1L1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.86 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene YME1L1
Gene Accession No. Q96TA2
Gene Alias ATP-dependent metalloprotease FtsH1; ATP-dependent metalloprotease FtsH1 homolog; ATP-dependent metalloprotease YME1L1; ATP-dependent metalloprotease YME1L1-like protein; ATP-dependent zinc metalloprotease YME1L1; ATP-dependent zinc metalloprotease YME1l1-like protein; FTSH; FTSH1; MEG4; meg-4; PAMP; Presenilin-associated metalloprotease; UNQ1868/PRO4304; YME1 like 1 ATPase; YME1L; YME1L1; YME1-like 1 (S. cerevisiae); YME1-like 1 ATPase; YME1-like protein 1
Gene Symbols YME1L1
Host Species Rabbit
Immunogen Recombinant Human ATP-dependent zinc metalloprotease YME1L1 protein (1-240AA).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10730
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.