Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

YME1L1 Polyclonal Antibody, CoraLite594, Proteintech
SDP

Catalog No. 502332725 Shop All Proteintech Group Inc Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-233-2725 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-233-2725 Supplier Proteintech Group Inc Supplier No. CL59411510100UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

YME1L1 Polyclonal antibody specifically detects YME1L1 in Human, Mouse, Rat samples. It is validated for Immunocytochemistry

Specifications

Antigen YME1L1
Applications Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate CoraLite 594
Formulation PBS with 0.5% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene Accession No. Q96TA2
Gene Alias FTSH; FTSH1; MEG4; PAMP; UNQ1868/PRO4304; YME1L; YME1L1
Host Species Rabbit
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10730
Target Species Mouse
Content And Storage -20° C, Avoid Freeze/Thaw Cycles, store in dark
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.