Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Ymer Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551563
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551563 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551563 Supplier Invitrogen™ Supplier No. PA551563
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QEKKDEDIAR LLQEKELQEE KKRKKHFPEF PATRAYADSY YYEDGGMKPR VMKEAVSTPS RMAHRDQEWY DAEIARKLQE EELLATQVDM RAAQVAQDEE IARLLMAEEK KAYKKAKERE KSSLDKRKQD PEWKPKTAKA ANSKSKESDE Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 81%.

This protein is a bifunctional mRNA-capping enzyme exhibiting RNA 5'-triphosphatase activity in the N-terminal part and mRNA guanylyltransferase activity in the C-terminal part. Catalyzes the first two steps of cap formation: by removing the gamma-phosphate from the 5'-triphosphate end of nascent mRNA to yield a diphosphate end, and by transferring the gmp moiety of GTP to the 5'-diphosphate terminus.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ymer
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Ccdc50
Gene Accession No. Q810U0, Q810U5, Q8IVM0
Gene Alias C3orf6; C3orf6h; Ccdc50; coiled-coil domain containing 50; coiled-coil domain-containing protein 50; DFNA44; Protein Ymer; putative C3orf6 protein homolog; YMER; Ymer protein
Gene Symbols Ccdc50
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human Ymer. Recombinant protein control fragment (Product #RP-95927).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 152137, 288022, 67501
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.