Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ YOD1 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA535042

Catalog No. PIPA535042


Only null left
Add to Cart

Description

Description

This antibody is predicted to react with chicken and zebrafish based on sequence homology.

Deubiquitinating enzymes (DUBs) are proteases that specifically cleave ubiquitin linkages, negating the action of ubiquitin ligases. DUBA8 belongs to a DUB subfamily characterized by an ovarian tumor (OTU) domain.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

YOD1
Polyclonal
Unconjugated
YOD1
9930028C20Rik; DUBA8; DUBA-8; fe01a10; HIN7; HIN-7; HIV-1-induced protease 7; HIV-induced protein-7-like protease; hsHIN7; hypothetical protein LOC550411; OTU domain containing 2; OTU domain-containing protein 2; OTUD2; PRO0907; Ubiquitin thioesterase OTU1; wu:fe01a10; Yod1; YOD1 deubiquitinase; YOD1 OTU deubiquinating enzyme 1 homolog; YOD1 OTU deubiquinating enzyme 1 homolog ( yeast); YOD1 OTU deubiquitinating enzyme 1 homologue; YOD1 OTU deubiquitinating enzyme 1 homologue (S. cerevisiae); zgc:112182
Rabbit
Antigen Affinity Chromatography, Protein A
RUO
55432
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
0.5 mg/mL
PBS with 0.09% sodium azide; pH 7.4
Q5VVQ6
YOD1
KLH conjugated synthetic peptide between 319-347 amino acids from the C-terminal region of human YOD1.
400 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.