Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ YOD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA589126
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA589126 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA589126 Supplier Invitrogen™ Supplier No. PA589126
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MFGPAKGRHF GVHPAPGFPG GVSQQAAGTK AGPAGAWPVG SRTDTMWRLR CKAKDGTHVL QGLSSRTRVR ELQGQIAAIT GIAPGGQRIL VGYPPECLDL SNGDTILEDL PIQSGDMLII EEDQTRPRSS PAFTKRGASS YVRETLPVLT RTVVPADNSC LFTSVYYVVE GGVLNPACAP EMRRLIAQIV ASDPDFYSEA ILGKTNQEYC DWIKRDDTWG GAIEISILSK FYQCEICVVD TQTVRIDRFG EDAGYTKRVL LIYDGIHYDP LQRNFPDPDT PPLTIFSSND DIVLVQALEL ADEARRRRQF TDVNRFTLRC MVCQKGLTGQ AEAREHAKET GHTNFGEV; Positive Samples: Mouse brain.

Deubiquitinating enzymes (DUBs) are proteases that specifically cleave ubiquitin linkages, negating the action of ubiquitin ligases. DUBA8 belongs to a DUB subfamily characterized by an ovarian tumor (OTU) domain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen YOD1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.88 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene YOD1
Gene Accession No. Q5VVQ6, Q8CB27
Gene Alias 9930028C20Rik; DUBA8; DUBA-8; fe01a10; HIN7; HIN-7; HIV-1-induced protease 7; HIV-induced protein-7-like protease; hsHIN7; hypothetical protein LOC550411; OTU domain containing 2; OTU domain-containing protein 2; OTUD2; PRO0907; Ubiquitin thioesterase OTU1; wu:fe01a10; Yod1; YOD1 deubiquitinase; YOD1 OTU deubiquinating enzyme 1 homolog; YOD1 OTU deubiquinating enzyme 1 homolog ( yeast); YOD1 OTU deubiquitinating enzyme 1 homologue; YOD1 OTU deubiquitinating enzyme 1 homologue (S. cerevisiae); zgc:112182
Gene Symbols YOD1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-348 of human YOD1 (NP_0610363).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 226418, 55432
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.