Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Yoda 1
GREENER_CHOICE

Catalog No. 558650
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
558650 50 mg
558610 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 558650 Supplier Tocris Bioscience™ Supplier No. 5586/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

Piezo1 channel activator

Yoda 1 is a selective activator of mouse and human mechanosensitive channel piezo1. In HEK cells, Yoda 1 slows the inactivation phase of transient currents, sensitizes Piezo1 to activation by pressure, and partially activates channels in the absence of external pressure. In MDCK cells, Yoda 1 induces Ca2+-dependent chromatin hypercondensation and alters Ca2+-dependent myosin contractility, leading to nuclear shrinkage in cells. In red blood cells from sickle cell anemia (SCA) patients, Yoda 1 increases intracellular Ca2+ and phosphatidylserine exposure, which leads to KCa3.1 channel mediated Ca2+ influx and K+ and water efflux, causing shrinkage of the cells.

Specifications

Chemical Name or Material 2-[5-[[(2,6-Dichlorophenyl)methyl]thio]-1,3,4-thiadiazol-2-yl]pyrazine
CAS 448947-81-7
Quantity 50 mg
Target Other Calcium Channel Activators
Molecular Formula C13H8Cl2N4S2
Formula Weight 355.27
Purity >99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.