Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Yohimbine hydrochloride
GREENER_CHOICESDP

Catalog No. 11271G Shop All Novus Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-271-G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 11-271-G Supplier R&D Systems™ Supplier No. 1127/1G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Selective α2 antagonist

α2-adrenoceptor antagonist (pKi values are 8.52, 8.00 and 9.17 for human α2A, α2B and α2C receptors respectively).

Diastereomer also available.

Chemical Identifiers

CAS 65-19-0
Molecular Formula C21H27ClN2O3

Specifications

Quantity 1 g
Product Line Tocris
Formula Weight 390.91
Purity >98%
Molecular Weight (g/mol) 390.91
Chemical Name or Material Yohimbine hydrochloride
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.