Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Youngstown Barrel & Drum Privacy Shelter for Ultra-Decon Decks™

Click to view available options
Capacity (English):
110 gal.
Description
- Vinyl coated nylon liner
- Waterproof
- White
Specifications
Specifications
Color | White |
Capacity (English) | 110 gal. |
Capacity (Metric) | 484.5 L |
Material | Nylon, Vinyl |
Type | Ultra-Decon Deck™ Optional Privacy Shelter |
For Use With (Application) | Tactical model and hospital model decontamination deck |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction