Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Youngstown Barrel & Drum HardTop Cover
SDP

Catalog No. 191304700
Click to view available options
Material:
Polyethylene

Lockable for indoor and outdoor use.

HardTop Cover only. Used with the Ultra Spill Colletion Collector System. Lockable for indoor and outdoor use.

Specifications

Color Yellow
Material Polyethylene
Type Hard Top Cover
For Use With (Application) Lockable usage, indoor and outdoor use
For Use With (Equipment) Ultra spill collection collector system
Model Collector System
Packaging Quantity 1 Ea.
Weight (English) 50 lb.
Weight (Metric) 22.68 kg
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.